Learn More
Med Vet International Jorvet Hypodermic Needle, 20g x 1-1/2in., J0174AD, 12/box
Catalog No. 5000581626
Encompass_Preferred
Jorvet 20g x 1-1/2in. Hypodermic needle is made of stainless steel, has a round hub, and is packaged in a cardboard box or clear plastic case. This product contains 12 needles.
Catalog No. 50-005-81626 Supplier Med Vet International Supplier No. JOR174AD
Jorvet 20g x 1-1/2in. Hypodermic needle is made of stainless steel, has a round hub, and is packaged in a cardboard box or clear plastic case. This product contains 12 needles.