Learn More
Med Vet International Jorvet Infusable 1 Liter Pressure Infusor, J0320P
Catalog No. 5000585611
Encompass_Preferred
Pressure infuser for collapsible IV bags. IV bag fits inside mesh and nylon bag. A rubber hand pump attached to a 3-way stop cock allows bag to be filled with air, which exerts pressure on the IV fluid bag.
Catalog No. 50-005-85611 Supplier Med Vet International Supplier No. JOR320P
Pressure infuser for collapsible IV bags. IV bag fits inside mesh and nylon bag. A rubber hand pump attached to a 3-way stop cock allows bag to be filled with air, which exerts pressure on the IV fluid bag.