Learn More
Med Vet International Jorvet Intraosseous Infusion Needle, 18g x 3.5cm., J0116Q, Each
Catalog No. 5000581467
Encompass_Preferred
Jorvet bone marrow intraoseous needle has a diamater of 18g and a length of 3.5cm. Using intraosseous infusion is an excellent option when vascular access is difficult. Using this method is mainly used in small animals and birds. The most common site of placement is the proximal femur and the point of entry for the canula is thre trochanteric fossa. This method is for short time use and an IV catheter should be placed as soon as possible. NY, CT, IL, NJ, NH, DE and MN restrict the sale of syringes and needles to licensed professionals and entities. By proceeding to purchase through our website, you acknowledge that you are over 18 years of age.
Catalog No. 50-005-81467 Supplier Med Vet International Supplier No. JOR116Q
Jorvet bone marrow intraoseous needle has a diamater of 18g and a length of 3.5cm. Using intraosseous infusion is an excellent option when vascular access is difficult. Using this method is mainly used in small animals and birds. The most common site of placement is the proximal femur and the point of entry for the canula is thre trochanteric fossa. This method is for short time use and an IV catheter should be placed as soon as possible. NY, CT, IL, NJ, NH, DE and MN restrict the sale of syringes and needles to licensed professionals and entities. By proceeding to purchase through our website, you acknowledge that you are over 18 years of age.