Learn More
Med Vet International JorVet IV Extension Set w/Two Y Sites, J1039A
Catalog No. 5000583188
Encompass_Preferred
Two "Y" sites 32" length Two slide clamps Male spin luer lock distal end Female luer lock proximal end
Catalog No. 50-005-83188 Supplier Med Vet International Supplier No. JOR1039A
Two "Y" sites 32" length Two slide clamps Male spin luer lock distal end Female luer lock proximal end