Learn More
Med Vet International Jorvet IV Holder w/ 4 Ring Holder, J0833H
Supplier:  Med Vet International JOR833H
Encompass_Preferred
This simple system puts a moveable IV bag holder on a track that is suspended from the ceiling. The IV holder can be positioned above cages, treatment area, or the surgery table. When not in use, it is simply retracted upward and pushed off to the side.Track can be installed into any type of ceiling.Lightweight aluminum track comes in any length sections; can be trimmed with hacksaw if needed.Four-wheel carrier runs in track. It is locked into position by turning it 360°. A pendant ring is attached to carrier.IV holder attached to carrier. it has 4 ram’s horns at 90° angle. They each can hold an IV bag. It extends and retracts at the push of a button.Oval sections of track are also available.IV holder for four bags (JOR833h)Four-wheel carrier with ring (JOR833c)Two end caps for track (JOR833e)
Catalog No. 50-005-85023