Learn More
Med Vet International Jorvet Lacrimal Needle, 23g, Straight, J0156A
Catalog No. 5000583043
Encompass_Preferred
Jorvet 23g lacrimal needle has a straight special tip for easy insertion into the lacrimal duct. It has a Luer mount so that it fits on any syringe. This needle is made of stainless steel, has an olive tip, and mesures 0.60mm x 30mm.
Catalog No. 50-005-83043 Supplier Med Vet International Supplier No. JOR156A
Jorvet 23g lacrimal needle has a straight special tip for easy insertion into the lacrimal duct. It has a Luer mount so that it fits on any syringe. This needle is made of stainless steel, has an olive tip, and mesures 0.60mm x 30mm.