Learn More
Med Vet International Jorvet Large Bore IV Set, J0400X
Catalog No. 5000582783
Encompass_Preferred
The JorVet large-bore IV set is a simple design for the rapid administration of solutions to large animals. Twice the diameter of conventional IV sets, the large bore increases the flow rate by a factor of 16 over standard human IV sets. Sterile.
Catalog No. 50-005-82783 Supplier Med Vet International Supplier No. JOR400X
The JorVet large-bore IV set is a simple design for the rapid administration of solutions to large animals. Twice the diameter of conventional IV sets, the large bore increases the flow rate by a factor of 16 over standard human IV sets. Sterile.