Learn More
Med Vet International Jorvet Michele Trephine, 7.5in, J0229, Each
Catalog No. 5000584377
Encompass_Preferred
Perfect instrument for a bone biopsy or drilling a small hole into the sinus. 6mm head. German-made. Stainless steel. Autoclavable. 7 1/2".
Catalog No. 50-005-84377 Supplier Med Vet International Supplier No. JOR229
Perfect instrument for a bone biopsy or drilling a small hole into the sinus. 6mm head. German-made. Stainless steel. Autoclavable. 7 1/2".