Learn More
Med Vet International Jorvet Needle, 23g x 1in, Blunt End, J0156D, Each
Supplier:  Med Vet International JOR156D
Encompass_Preferred
Jorvet 30g x 1in. Blunt tip needle without olive tip, has a stainless steel shaft and luer lock fitting.
Catalog No. 50-005-83024