Learn More
Med Vet International Jorvet Needle, 25g x 1in., Blunt Tip, J1032K, 5/pack
Catalog No. 5000583527
Encompass_Preferred
Jorvet 25g x 1in. blunt tip needles have polished straight stainless cannula with a polypropylene color-coded hub. These needles have a wide range of applications around any veterinary clinic including: Anal Sac Irrigation Wound Flushing Lacrimal Duct Flush Male Cat urethral flush Feeding Needle NY, CT, IL, NJ, NH, DE and MN restrict the sale of syringes and needles to licensed professionals and entities. By proceeding to purchase through our website, you acknowledge that you are over 18 years of age.
Catalog No. 50-005-83527 Supplier Med Vet International Supplier No. JOR1032K
Jorvet 25g x 1in. blunt tip needles have polished straight stainless cannula with a polypropylene color-coded hub. These needles have a wide range of applications around any veterinary clinic including: Anal Sac Irrigation Wound Flushing Lacrimal Duct Flush Male Cat urethral flush Feeding Needle NY, CT, IL, NJ, NH, DE and MN restrict the sale of syringes and needles to licensed professionals and entities. By proceeding to purchase through our website, you acknowledge that you are over 18 years of age.