Learn More
Med Vet International Jorvet OB Calving Strap, tubular Nylon Strap and D Ring, 60in., J0024XL
Catalog No. 5000585043
Encompass_Preferred
High quality tubular nylon strap with "D" ring. This strap is less traumatic to the unborn calf than steel chains. Royal blue. 1"W.
Catalog No. 50-005-85043 Supplier Med Vet International Supplier No. JOR24XL
High quality tubular nylon strap with "D" ring. This strap is less traumatic to the unborn calf than steel chains. Royal blue. 1"W.