Learn More
Med Vet International Jorvet Polypro Needle, 20g x 1-1/2in., Blunt Tip, J1033D, 5/pack
Catalog No. 5000583206
Encompass_Preferred
Jorvet 20g x 1-1/2in. polypropylene blunt tip needles have a polypropylene burr free shaft with a poly hubs. These needles have a wide range of applications around any veterinary clinic including: Anal Sac Irrigation Wound Flushing Lacrimal Duct Flush Male Cat urethral flush Feeding Needle NY, CT, IL, NJ, NH, DE and MN restrict the sale of syringes and needles to licensed professionals and entities. By proceeding to purchase through our website, you acknowledge that you are over 18 years of age.
Catalog No. 50-005-83206 Supplier Med Vet International Supplier No. JOR1033D
Jorvet 20g x 1-1/2in. polypropylene blunt tip needles have a polypropylene burr free shaft with a poly hubs. These needles have a wide range of applications around any veterinary clinic including: Anal Sac Irrigation Wound Flushing Lacrimal Duct Flush Male Cat urethral flush Feeding Needle NY, CT, IL, NJ, NH, DE and MN restrict the sale of syringes and needles to licensed professionals and entities. By proceeding to purchase through our website, you acknowledge that you are over 18 years of age.