Learn More
Med Vet International Jorvet Rosenthal Bone Biopsy Needle, 16g x 1-5/16in, J0116RB, Each
Catalog No. 5000581071
Encompass_Preferred
The Jorvet Rosentahal Bone marrow biopsy needle has a 16g diamater and a length of 1-5/16in. Thise needle has a large spool luer lock hub, a sharp pointed cannula, and a fitted stylet with matched bevel point. This needle is autoclavable. NY, CT, IL, NJ, NH, DE and MN restrict the sale of syringes and needles to licensed professionals and entities. By proceeding to purchase through our website, you acknowledge that you are over 18 years of age.
Catalog No. 50-005-81071 Supplier Med Vet International Supplier No. JOR116RB
The Jorvet Rosentahal Bone marrow biopsy needle has a 16g diamater and a length of 1-5/16in. Thise needle has a large spool luer lock hub, a sharp pointed cannula, and a fitted stylet with matched bevel point. This needle is autoclavable. NY, CT, IL, NJ, NH, DE and MN restrict the sale of syringes and needles to licensed professionals and entities. By proceeding to purchase through our website, you acknowledge that you are over 18 years of age.