Learn More
Med Vet International Jorvet Safety Stick Pole Syringe, J0482B, 4ft, 3cc, Each
Catalog No. 5000584562
Encompass_Preferred
Jorvet 4ft safety stick pole syringe allows the user to obtain a proper needle stick and push the dosage plunger on the operator ended. This product uses a standard 3cc syringe. The pole is constructed of aircraft aluminum alloy and stainless steel that cannot be damaged by even the most powerful animal bite and has a black anodized satin finish.
Catalog No. 50-005-84562 Supplier Med Vet International Supplier No. JOR482B
Jorvet 4ft safety stick pole syringe allows the user to obtain a proper needle stick and push the dosage plunger on the operator ended. This product uses a standard 3cc syringe. The pole is constructed of aircraft aluminum alloy and stainless steel that cannot be damaged by even the most powerful animal bite and has a black anodized satin finish.