Learn More
Med Vet International Jorvet Safety Stick Pole Syringe, J0482C, 6ft, 3cc, Each
Catalog No. 5000585072
Encompass_Preferred
Jorvet 6ft safety stick pole syringe allows the user to obtain a proper needle stick and push the dosage plunger on the operator ended. This product uses a standard 3cc syringe. The pole is constructed of aircraft aluminum alloy and stainless steel that cannot be damaged by even the most powerful animal bite and has a black anodized satin finish.
Catalog No. 50-005-85072 Supplier Med Vet International Supplier No. JOR482C
Jorvet 6ft safety stick pole syringe allows the user to obtain a proper needle stick and push the dosage plunger on the operator ended. This product uses a standard 3cc syringe. The pole is constructed of aircraft aluminum alloy and stainless steel that cannot be damaged by even the most powerful animal bite and has a black anodized satin finish.