Learn More
Med Vet International Jorvet Semen Extender, without Antibiotics, J0124N, 10/pack
Catalog No. 5000584173
Encompass_Preferred
This is the all natural product of glucose and non-fat drymilk. It is recommended for all artificial insemination purposes. The semen may be cooled for extended storageperiods for transport. It may also be used for same day breeding on the farm with or without antibiotics. Each kit contains the component parts in a hermetically sealed pouch and a separate 150ml wide-mouth bottle of sterile water.
Catalog No. 50-005-84173 Supplier Med Vet International Supplier No. JOR124N
This is the all natural product of glucose and non-fat drymilk. It is recommended for all artificial insemination purposes. The semen may be cooled for extended storageperiods for transport. It may also be used for same day breeding on the farm with or without antibiotics. Each kit contains the component parts in a hermetically sealed pouch and a separate 150ml wide-mouth bottle of sterile water.