Learn More
Med Vet International Jorvet Small Animal Blood Collection Set, Including Syringe & Stopcock, J0520AX, Each
Catalog No. 5000582296
Encompass_Preferred
The addition of these blood collection sets allows for easier and proper feline and small animal blood collection. They are a complete closed system that will get better blood component results, i.e., packed RBC or plasma. The bags are provided dry and anticoagulant (JOR522) will need to be added. The sets come in two basic formats: a set with 19g butterfly needle or a 60cc syringe with a 3-way stop cock.Small blood bag collection set.100 dry (blood) collection bag19g butterfly needle with tubingcomplete directions
Catalog No. 50-005-82296 Supplier Med Vet International Supplier No. JOR520AX
The addition of these blood collection sets allows for easier and proper feline and small animal blood collection. They are a complete closed system that will get better blood component results, i.e., packed RBC or plasma. The bags are provided dry and anticoagulant (JOR522) will need to be added. The sets come in two basic formats: a set with 19g butterfly needle or a 60cc syringe with a 3-way stop cock.Small blood bag collection set.100 dry (blood) collection bag19g butterfly needle with tubingcomplete directions