Learn More
Med Vet International Jorvet Smaller Barbed Connector, J0282D4
Catalog No. 5000585573
Encompass_Preferred
Smaller barbed connector with female luer lock. 2.8mm-7mm.
Catalog No. 50-005-85573 Supplier Med Vet International Supplier No. JOR282D4
Smaller barbed connector with female luer lock. 2.8mm-7mm.