Learn More
Med Vet International Jorvet Spring Infusor Tubing 10mL/30 Min, J0831B, 10 Pack
Catalog No. 5000582217
Encompass_Preferred
A simple reusable spring-driver pump used for continuous IV infusions over time, requires no batteries or electricity. The pump is matched with various special sized tubing to determine actual flow rate. Uses include adminstering oncology and anesthetic drugs, and IV fluids to very small patients. ONLY TUBINGFeatures:Large variety of flow rates available (e.g., 10ml, 5 minutes to 8 hours)Compact and very portableReusableNo programming requiredNot gravity dependent
Catalog No. 50-005-82217 Supplier Med Vet International Supplier No. JOR831B
A simple reusable spring-driver pump used for continuous IV infusions over time, requires no batteries or electricity. The pump is matched with various special sized tubing to determine actual flow rate. Uses include adminstering oncology and anesthetic drugs, and IV fluids to very small patients. ONLY TUBINGFeatures:Large variety of flow rates available (e.g., 10ml, 5 minutes to 8 hours)Compact and very portableReusableNo programming requiredNot gravity dependent