Learn More
Med Vet International Jorvet Thunderbolt Hypodermic Needle, 14g x 1.5in, J0958, 100/Box
Catalog No. 5000582421
Encompass_Preferred
Jorvet 14g x 1-1/2in. Thunderbolt Hypodermic needles have an aluminum hub with stainless steel needle cannulas. Designed for the demands of injecting large animlas, the needles are razor sharp and individually packaged in their own disposable 2 piece cartridges. Each needle size is color coded by gauge and marked with needle guage and length.
Catalog No. 50-005-82421 Supplier Med Vet International Supplier No. JOR958
Jorvet 14g x 1-1/2in. Thunderbolt Hypodermic needles have an aluminum hub with stainless steel needle cannulas. Designed for the demands of injecting large animlas, the needles are razor sharp and individually packaged in their own disposable 2 piece cartridges. Each needle size is color coded by gauge and marked with needle guage and length.