Learn More
Med Vet International Jorvet Transfer Bag, 150 ml, J0520TA
Catalog No. 5000581995
Encompass_Preferred
Designed to receive blood components or plasma, transfer bags have 2 spike entry ports and a single transfer spike with tubing. Sterile. Do not contain anticoagulants.
Catalog No. 50-005-81995 Supplier Med Vet International Supplier No. JOR520TA
Designed to receive blood components or plasma, transfer bags have 2 spike entry ports and a single transfer spike with tubing. Sterile. Do not contain anticoagulants.