Learn More
Med Vet International Jorvet Transfer Bag, 600ml, J0520TC
Catalog No. 5000581996
Encompass_Preferred
Designed to receive blood components or plasma, transfer bags have 2 spike entry ports and a single transfer spike with tubing. Sterile. Do not contain anticoagulants.
Catalog No. 50-005-81996 Supplier Med Vet International Supplier No. JOR520TC
Designed to receive blood components or plasma, transfer bags have 2 spike entry ports and a single transfer spike with tubing. Sterile. Do not contain anticoagulants.