Learn More
Med Vet International Jorvet Transfer Needle, 14g x 3in., J0173T
Catalog No. 5000582836
Encompass_Preferred
Has a concave metal plate where the finger can steady the transfer needle during mixing. Stainless steel. 14g x 3". Packaged in plastic vial.
Catalog No. 50-005-82836 Supplier Med Vet International Supplier No. JOR173T
Has a concave metal plate where the finger can steady the transfer needle during mixing. Stainless steel. 14g x 3". Packaged in plastic vial.