Learn More
Med Vet International Jorvet Tru-Core II 14g x 16cm Auto Biopsy Needle, J0528G
Catalog No. 5000581985
Encompass_Preferred
Jorvet 14g x 16cm Tru-Core II auto biopsy needle is the next generation of soft tissue biopsy instruemtns. In Human medicine Tru-Core II is widely used because of the ease of use, more represenative samples. These needles have an echogenic tip for use with ultrasounds.
Catalog No. 50-005-81985 Supplier Med Vet International Supplier No. JOR528G
Jorvet 14g x 16cm Tru-Core II auto biopsy needle is the next generation of soft tissue biopsy instruemtns. In Human medicine Tru-Core II is widely used because of the ease of use, more represenative samples. These needles have an echogenic tip for use with ultrasounds.