Learn More
Med Vet International Jorvet Tru-Core II 16g x 10cm Auto Biopsy Needle, J0528H
Catalog No. 5000582310
Encompass_Preferred
Jorvet 16g x 10cm Tru-Core II auto biopsy needle is the next generation of soft tissue biopsy instruemtns. In Human medicine Tru-Core II is widely used because of the ease of use, more represenative samples. These needles have an echogenic tip for use with ultrasounds.
Catalog No. 50-005-82310 Supplier Med Vet International Supplier No. JOR528H
Jorvet 16g x 10cm Tru-Core II auto biopsy needle is the next generation of soft tissue biopsy instruemtns. In Human medicine Tru-Core II is widely used because of the ease of use, more represenative samples. These needles have an echogenic tip for use with ultrasounds.