Learn More
Med Vet International Jorvet Tru Cut Biopsy Needle, 14g x 15cm, J0118A, Each
Catalog No. 5000581094
Encompass_Preferred
The Jorvet 14g Tru-Cut Biopsy Needle, is long enough to reach any biopsy site. It has a 1mm x 20mm specimen chamber and has a 15cm shaft.
Catalog No. 50-005-81094 Supplier Med Vet International Supplier No. JOR118A
The Jorvet 14g Tru-Cut Biopsy Needle, is long enough to reach any biopsy site. It has a 1mm x 20mm specimen chamber and has a 15cm shaft.