Learn More
Med Vet International Jorvet Ultrasite IV Adapter, J0463Q
Catalog No. 5000581612
Encompass_Preferred
Allows needle free "syringing" from rubber multi-dose vial bottles without the risk of contamination.
Catalog No. 50-005-81612 Supplier Med Vet International Supplier No. JOR463Q
Allows needle free "syringing" from rubber multi-dose vial bottles without the risk of contamination.