Learn More
Med Vet International JorVet Veterinary Surgical Drape, 24" x 32", Opening: 2" x 1", J0256B
Catalog No. 5000582554
Encompass_Preferred
50% cotton, 50% polyester surgical grade textile Fenestrated drapes feature a moisture-proof barrier near the fenestration that repels body fluids Durable quality, may be safely washed and autoclaved frequently
Catalog No. 50-005-82554 Supplier Med Vet International Supplier No. JOR256B
50% cotton, 50% polyester surgical grade textile Fenestrated drapes feature a moisture-proof barrier near the fenestration that repels body fluids Durable quality, may be safely washed and autoclaved frequently