Learn More
Med Vet International JorVet Veterinary Surgical Leg Drape, 20" x 24", Opening: 6"x 9", J0256G
Catalog No. 5000582999
Encompass_Preferred
50% cotton, 50% polyester surgical grade textile Fenestrated drapes feature a moisture-proof barrier near the fenestration that repels body fluids Durable quality, may be safely washed and autoclaved frequently
Catalog No. 50-005-82999 Supplier Med Vet International Supplier No. JOR256G
50% cotton, 50% polyester surgical grade textile Fenestrated drapes feature a moisture-proof barrier near the fenestration that repels body fluids Durable quality, may be safely washed and autoclaved frequently