Learn More
Med Vet International JorVet Veterinary Surgical Leg Drape, 20" x 24", Opening: 6" x 9", J0256H
Catalog No. 5000583107
Encompass_Preferred
50% cotton, 50% polyester surgical grade textile Fenestrated drapes feature a moisture-proof barrier near the fenestration that repels body fluids Durable quality, may be safely washed and autoclaved frequently
Catalog No. 50-005-83107 Supplier Med Vet International Supplier No. JOR256H
50% cotton, 50% polyester surgical grade textile Fenestrated drapes feature a moisture-proof barrier near the fenestration that repels body fluids Durable quality, may be safely washed and autoclaved frequently