Learn More
Med Vet International Jorvet Winged IV Infusion Set, 19G x 12in, J0454A, Yellow, 100/Box
Catalog No. 5000581679
Encompass_Preferred
These popular sets are commonly known as Butterfly style IV sets. They are great for short term IV infusion or in difficult venipunctures. Tubing with female luer fitting is 12" long. A 3/4" razor sharp needle is attached to flexible colored coded wings. Priced very inexpensive. Sterile. 100/box.
Catalog No. 50-005-81679 Supplier Med Vet International Supplier No. JOR454A
These popular sets are commonly known as Butterfly style IV sets. They are great for short term IV infusion or in difficult venipunctures. Tubing with female luer fitting is 12" long. A 3/4" razor sharp needle is attached to flexible colored coded wings. Priced very inexpensive. Sterile. 100/box.