Learn More
Med Vet International JorvetTru Cut Biopsy Needle, 18g x 10cm, J0118D, Each
Catalog No. 5000581095
Encompass_Preferred
The Jorvet 18g Tru-Cut Biopsy Needle, is long enough to reach any biopsy site. It has a 1mm x 20mm specimen chamber and has a 10cm shaft.
Catalog No. 50-005-81095 Supplier Med Vet International Supplier No. JOR118D
The Jorvet 18g Tru-Cut Biopsy Needle, is long enough to reach any biopsy site. It has a 1mm x 20mm specimen chamber and has a 10cm shaft.