Learn More
TARGETMOL CHEMICALS INC JOSAMYCIN 100MG
SDP
Catalog No. 502576345
Encompass_Preferred
Also available in 25 mg 50 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Josamycin (EN-141) is a macrolide antibiotic exhibiting antimicrobial activity. The dissociation constant Kd from ribosome for Josamycin is 5.5 nM. purity: 99%
Catalog No. 50-257-6345 Supplier TARGETMOL CHEMICALS INC Supplier No. T7148100MG
Also available in 25 mg 50 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Josamycin (EN-141) is a macrolide antibiotic exhibiting antimicrobial activity. The dissociation constant Kd from ribosome for Josamycin is 5.5 nM. purity: 99%