Learn More
AdipoGen Josamycin
SDP
Catalog No. 502560623
Encompass_Preferred
Chemical. CAS 16846-24-5. Formula C42H69NO15. MW 827.99. Synthetic. A member of the leucomycin family of macrolide antibiotics produced by Streptomyces kitasatoensis. May overcome anticancer drug resistance by inhibiting the binding of vinblastine or cyclosporin A to P-glycoprotein Pgp. An antimicrobial against a wide variety of pathogens.
Catalog No. 50-256-0623 Supplier AdipoGen Supplier No. CDXJ0001M200
Chemical. CAS 16846-24-5. Formula C42H69NO15. MW 827.99. Synthetic. A member of the leucomycin family of macrolide antibiotics produced by Streptomyces kitasatoensis. May overcome anticancer drug resistance by inhibiting the binding of vinblastine or cyclosporin A to P-glycoprotein Pgp. An antimicrobial against a wide variety of pathogens.