Learn More
Medchemexpress LLC HY-B1920 25mg Medchemexpress, Josamycin CAS:16846-24-5 Purity:>98%
SDP
Catalog No. 501933039
Encompass_Preferred
Medchemexpress, HY-B1920 25mg Josamycin CAS:16846-24-5 Josamycin (EN-141) is a macrolide antibiotic exhibiting antimicrobial activity against a wide spectrum of pathogens, such as bacteria . The dissociation constant K d from ribosome for Josamycin is 5.5 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3039 Supplier Medchemexpress LLC Supplier No. HYB192025MG
Medchemexpress, HY-B1920 25mg Josamycin CAS:16846-24-5 Josamycin (EN-141) is a macrolide antibiotic exhibiting antimicrobial activity against a wide spectrum of pathogens, such as bacteria . The dissociation constant K d from ribosome for Josamycin is 5.5 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.