Learn More
AdipoGen Josamycin (50 mg)
SDP
Supplier:  AdipoGen CDXJ0001M050
Encompass_Preferred
Josamycin (50 mg), AdipoGen Life Sciences. Chemical. CAS: 16846-24-5. Formula: C42H69NO15. MW: 827.99. Synthetic. A member of the leucomycin family of macrolide antibiotics produced by Streptomyces kitasatoensis. May overcome anticancer drug resistance by inhibiting the binding of vinblastine or cyclosporin A to P-glycoprotein (Pgp). An antimicrobial against a wide variety of pathogens.
Catalog No. 50-149-0474