Learn More
Cayman Chemical Josamycin, Purity: ≥90%, CAS: 16846-24-5, 100 mg
SDP
Catalog No. 502990545
Encompass_Preferred
A macrolide antibiotic; active against clinical isolates of the Gram-positive aerobic bacteria (MIC50s = ≤0.39 µg/ml for all), as well as Gram-negative anaerobic bacteria at 6.25 µg/ml; increases survival in mouse models of systemic S. aureus, S. pyogenes, and S. pneumoniae infection (ED50s = 206.8, 205, and 86.7 mg/kg, respectively)
Catalog No. 50-299-0545 Supplier Cayman Chemical Supplier No. 29606100 MG
A macrolide antibiotic; active against clinical isolates of the Gram-positive aerobic bacteria (MIC50s = ≤0.39 µg/ml for all), as well as Gram-negative anaerobic bacteria at 6.25 µg/ml; increases survival in mouse models of systemic S. aureus, S. pyogenes, and S. pneumoniae infection (ED50s = 206.8, 205, and 86.7 mg/kg, respectively)