Learn More
Cayman Chemical Josamycn 50mg
SDP
Catalog No. 502990322
Encompass_Preferred
A peptide agonist of CaSR selective for CaSR over a panel of 33 receptors and ion channels as well as the norepinephrine transporter at 10 UM increases intracellular calcium levels in HEK293T cells expressing the human CaSR receptor (EC50 0 53 UM) inhibits parathyroid secretion from primary rat parathyroid cells (EC50 0 36 UM in the presence of calcium) decreases parathyroid hormone and calcium in plasma and serum respectively in a model of chronic renal insufficiency with secondary hyperthyroidism in nephrectomized rats fed high-phosphorus diet at 0 3 1 and 3 mg/kg
Catalog No. 50-299-0322 Supplier Cayman Chemical Supplier No. 2960650 MG
A peptide agonist of CaSR selective for CaSR over a panel of 33 receptors and ion channels as well as the norepinephrine transporter at 10 UM increases intracellular calcium levels in HEK293T cells expressing the human CaSR receptor (EC50 0 53 UM) inhibits parathyroid secretion from primary rat parathyroid cells (EC50 0 36 UM in the presence of calcium) decreases parathyroid hormone and calcium in plasma and serum respectively in a model of chronic renal insufficiency with secondary hyperthyroidism in nephrectomized rats fed high-phosphorus diet at 0 3 1 and 3 mg/kg