Learn More
Surgical Design™ Joseph Bone Saw
Catalog No. 21124642 Shop All Surgical Design Products
Change view
Click to view available options
Shape:
Bayonet, Left
Bayonet, Right
Straight
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Shape
21-124-642 Straight
21-124-643 Bayonet, Left
21-124-644 Bayonet, Right
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 21-124-642 Supplier Surgical Design™ Supplier No. ATPM021
Only null left
Add to Cart
Add to Cart
Joseph bone saw is designed for precise bone cutting during autopsy with control in confined anatomical spaces.
  • Joseph Bone Saw, 7-in. length, engineered for detailed bone cutting during autopsy and forensic examinations
  • Available in three configurations:Straight standard pattern for general bone sectioning
  • Bayonet Left offset design improves visibility and maneuverability on the left side of the body or angled cutting paths
  • Bayonet Right mirrored offset for right-sided access or complex anatomical angles
  • Fine, aggressive serrated blade delivers smooth, controlled cutting of ribs, small long bones, vertebrae, and cranial structures
  • Bayonet offset designs allow the hand to remain clear of the cutting line, enhancing precision in confined or deep spaces
  • Ideal for cranial, cervical, and thoracic dissection, especially around tight anatomical curves
  • Made from premium surgical-grade stainless steel for long life and resistance to corrosion in autopsy environments
  • Ergonomic handle provides a secure grip even in wet or high-debris conditions
  • Fully autoclavable and reusable, suitable for high-volume forensic and pathology labs
  • Designed for human autopsy, forensic pathology, medical examiner facilities, and necropsy work
  • Cranial opening and skull plate separation
  • Rib, vertebral, and small bone sectioning
  • Precise bone cuts in tight or angled regions (especially with bayonet variants)
  • Dissection around the neck, base of the skull, and thoracic inlet
  • Suitable for both human autopsy and veterinary necropsy
Specifications
Product Type Bone Saw
Certifications/Compliance ISO/GMP
Material Stainless Steel
Shape Straight
Sterility Non-sterile