Learn More
Surgical Design™ Joseph-Maltz Nasal Saw - Angular, Bayonet Left/Right
Catalog No. 21124647 Shop All Surgical Design Products
Change view
Click to view available options
Shape:
Bayonet, Left
Bayonet, Right
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Shape
21-124-647 Bayonet, Left
21-124-648 Bayonet, Right
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-124-647 Supplier Surgical Design™ Supplier No. ATPM026
Only null left
Add to Cart
Add to Cart
Joseph-Maltz angular nasal saw is designed for precise bone cutting in tight anatomical spaces during autopsy.
  • Joseph-Maltz Nasal Saw, available in Angular Left Bayonet and Angular Right Bayonet configurations
  • Designed for precise bone cutting in confined regions, especially the nasal cavity and midface
  • Bayonet offset provides unobstructed visibility and improved maneuverability during delicate facial bone dissection
  • Ideal for sectioning nasal bones, maxilla edges, orbital margins, and anterior skull structures during autopsy
  • Angular blade shape allows controlled access to deep or narrow anatomical areas
  • Suitable for forensic reconstruction, detailed facial examination, and access to sinus structures
  • Made from surgical-grade stainless steel for long-lasting strength and corrosion resistance
  • Ergonomic handle ensures a steady grip in wet or fluid-rich postmortem environments
  • Fully reusable and autoclavable, built for routine use in medical examiner and pathology settings
  • Removal or sectioning of nasal bones and anterior facial structures
  • Accessing and evaluating sinus cavities
  • Precise cuts for facial reconstruction in forensic cases
  • Controlled bone work around orbital rims and frontal processes
Specifications
Product Type Nasal Bone Saw
Certifications/Compliance ISO/GMP
Material Stainless Steel
Shape Bayonet, Left
Sterility Non-sterile