Learn More
Strem, An Ascensus Company JosiphosPalladacycleG398%500mg
SDP
Supplier:  Strem, An Ascensus Company 460353500MG
Encompass
CAS# 1702311-34-9. 500mg. Methanesulfonato{(R)-(-)-1-[(S)-2-(dicyclohexylphosphino)ferrocenyl]ethyldi-t-butylphosphine}(2'-amino-1,1'-biphenyl-2-yl)palladium(II), min. 98% [Josiphos Palladacycle Gen. 3]. MFCD27978424. Molecular Weight: 924.28. Molecular Formula: C45H65FeNO3P2PdS. Color/form: red-orange solid Strem# 46-0353. http://www.strem.com/catalog/v/46-0353/
Catalog No. 50-203-4167