Learn More
Enterprise Technology Solutions JoySuds Detergent,Powder,Manual,Multipurpose,50 lb,WE
SDP
Catalog No. 5000810880
Encompass_Preferred
Cream Suds Food Processing Equipment Presoak and Cleaner has your commercial kitchen needs covered with a professional formula built to handle tough jobs and leave your facility smelling great. Powder-based detergent cuts through heavy grease and soaks baked-on foods on pots and pans. It is also effective on a variety of other washable surfaces: stainless steel, floors, walls and countertops. Simply dilute the double-concentrated detergent with water and spray on surfaces for "clean-in-place" items.
Catalog No. 50-008-10880 Supplier Enterprise Technology Solutions Supplier No. JYS43612
Cream Suds Food Processing Equipment Presoak and Cleaner has your commercial kitchen needs covered with a professional formula built to handle tough jobs and leave your facility smelling great. Powder-based detergent cuts through heavy grease and soaks baked-on foods on pots and pans. It is also effective on a variety of other washable surfaces: stainless steel, floors, walls and countertops. Simply dilute the double-concentrated detergent with water and spray on surfaces for "clean-in-place" items.