Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JP-5 Military Fuel Solution, 5000μg/mL in Dichloromethane, Ultra Scientific

Catalog No. USRGO6911
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-RGO-691-1 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-RGO-691-1 Supplier Agilent Technologies Supplier No. RGO6911
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Specifications

Chemical Name or Material JP-5 Military Fuel Solution
Name Note 5000μg/mL (Methylene chloride)
Physical Form Liquid
Molecular Formula CH2Cl2
MDL Number MFCD00000881
Color Colorless
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
SMILES ClCCl
IUPAC Name dichloromethane
Molecular Weight (g/mol) 84.93
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 5000μg/mL (Methylene chloride)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.