Learn More
TARGETMOL CHEMICALS INC JP1302 dihydrochloride 10MG
SDP
Catalog No. 502878178
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. JP1302 dihydrochloride is a selective, high affinity antagonist of the alpha2C-adrenoceptor (alpha2C-adrenoceptor), with a Ki (binding affinity) value of 28 nM and a Kb value (antagonist activity) of 16 nM. Purity 97.83%
Catalog No. 50-287-8178 Supplier TARGETMOL CHEMICALS INC Supplier No. T1172610MG
Also available in 1 mL, 1 mg, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. JP1302 dihydrochloride is a selective, high affinity antagonist of the alpha2C-adrenoceptor (alpha2C-adrenoceptor), with a Ki (binding affinity) value of 28 nM and a Kb value (antagonist activity) of 16 nM. Purity 97.83%