Learn More
Medchemexpress LLC JP1302 dihydrochloride | 1259314-65-2 | MFCD10565599 | >98.0% | 441.40 | C24H26Cl2N4 | 5 MG
SDP
Catalog No. 5000247337
Encompass_Preferred
JP1302 dihydrochloride is a research-grade small molecule antagonist selective for the α2C-adrenoceptor, supplied as the dihydrochloride salt for improved solubility. It has reported high affinity for human α2C receptors (Kb ≈ 16 nM; Ki ≈ 28 nM) and is used in preclinical studies of neuropsychiatric and renal dysfunction models. Intended for laboratory research use only.
  • High affinity α2C-adrenoceptor antagonist (Kb ≈ 16 nM; Ki ≈ 28 nM).
  • Dihydrochloride salt form for improved stability and solubility.
  • Molecular formula C24H26Cl2N4; molecular weight 441.40.
  • Supplied in small research quantities suitable for analytical and preclinical use.
  • For laboratory research only; not for human or veterinary use.
Catalog No. 50-002-47337 Supplier Medchemexpress LLC Supplier No. HY1032135MG
JP1302 dihydrochloride is a research-grade small molecule antagonist selective for the α2C-adrenoceptor, supplied as the dihydrochloride salt for improved solubility. It has reported high affinity for human α2C receptors (Kb ≈ 16 nM; Ki ≈ 28 nM) and is used in preclinical studies of neuropsychiatric and renal dysfunction models. Intended for laboratory research use only.
  • High affinity α2C-adrenoceptor antagonist (Kb ≈ 16 nM; Ki ≈ 28 nM).
  • Dihydrochloride salt form for improved stability and solubility.
  • Molecular formula C24H26Cl2N4; molecular weight 441.40.
  • Supplied in small research quantities suitable for analytical and preclinical use.
  • For laboratory research only; not for human or veterinary use.