Learn More
Medchemexpress LLC JP1302 dihydrochloride | 1259314-65-2 | MFCD10565599 | 99.9% | 441.40 g/mol | C24H26Cl2N4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10321310MG
Encompass_Preferred
JP1302 dihydrochloride is a dihydrochloride salt small molecule antagonist of the human α2C-adrenoceptor supplied as a solid for research use.
  • High purity (99.9%).
  • Potent, selective α2C-adrenoceptor antagonist (Kb 16 nM; Ki 28 nM).
  • Brown to reddish brown solid appearance.
  • Soluble in water and DMSO at research-relevant concentrations (see datasheet for details).
  • Store sealed away from moisture; in solvent store at -80°C for up to 6 months or -20°C for up to 1 month.
Catalog No. 50-002-46296