Learn More
Abcam JP1302 dihydrochloride, alpha2C antagonist, 10MG
Supplier:  Abcam AB14112910MG
Encompass_Preferred
MW 441.4 g/mol, Purity >99%. Potent and highly selective α₂C antagonist (Kb values for human subtypes are 28 (α₂C), 1470 (α₂B), 1700 (α₂D) and 3150 nM (α₂A)). Shows antidepressant and antipsychotic effects. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6872