Learn More
Aviva Systems Biology Corp JPH1 Antibody - C-terminal region (ARP49533_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49533_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against JPH1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human JPH1. Alternate names: DKFZp762L0313, JP-1, JP1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60105