Learn More
Aviva Systems Biology Corp JPH2 Antibody - middle region (ARP51358_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51358_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against JPH2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human JPH2. Alternate names: FLJ40969, JP-2, JP2, CMH17ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60560