Learn More
Medchemexpress LLC Jpm-oet | 262381-84-0 | 99.0% | 392.45 g/mol | C20H28N2O6 | 1 ML
SDP
Catalog No. 5000246191
Encompass_Preferred
JPM-OEt is a covalent, irreversible inhibitor of the cysteine cathepsin family used in biochemical and cellular research to probe protease activity and to investigate antitumor effects. It is supplied either as a ready-to-use 10 mM solution in DMSO or as solid material for formulation and storage.
  • Covalent, irreversible inhibition of cysteine cathepsins.
  • Useful for biochemical and cellular assays probing protease function.
  • Reported antitumor activity in preclinical studies.
  • Available as 10 mM solution in DMSO and multiple solid sizes.
  • Stable when stored frozen in solution; solid material stored refrigerated and protected from light.
Catalog No. 50-002-46191 Supplier Medchemexpress LLC Supplier No. HY10208710MM 1ML
JPM-OEt is a covalent, irreversible inhibitor of the cysteine cathepsin family used in biochemical and cellular research to probe protease activity and to investigate antitumor effects. It is supplied either as a ready-to-use 10 mM solution in DMSO or as solid material for formulation and storage.
  • Covalent, irreversible inhibition of cysteine cathepsins.
  • Useful for biochemical and cellular assays probing protease function.
  • Reported antitumor activity in preclinical studies.
  • Available as 10 mM solution in DMSO and multiple solid sizes.
  • Stable when stored frozen in solution; solid material stored refrigerated and protected from light.